Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177368.4 | NP_796342.2 | 10 - 11 | 3003 | 64 | |
GenBank mRNA | BC034657.1 | - | 3 - 4 | 224 | 64 | |
BC090655.1 | - | 10 - 11 | 2365 | 64 | ||
BC092226.1 | - | 10 - 11 | 2959 | 6852756 | 64 | |
BC138362.1 | - | 10 - 11 | 2734 | 8861382 | 64 | |
BC138363.1 | - | 10 - 11 | 2734 | 8861383 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |