Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145439.2 | NP_663414.3 | 19 - 20 | 2564 | 88 | |
GenBank mRNA | AK090179.1 | - | 19 - 20 | 2519 | 88 | |
AK191662.1 | - | 1 - 2 | 75 | 88 | ||
AK191892.1 | - | 1 - 2 | 75 | 88 | ||
AK191934.1 | - | 1 - 2 | 75 | 88 | ||
AY236497.1 | - | 19 - 20 | 88 | |||
AY263158.1 | - | 19 - 20 | 2489 | 88 | ||
BC004840.1 | - | 19 - 20 | 2489 | 2648849 | 88 | |
BC005510.1 | - | 12 - 13 | 1509 | 88 | ||
BC006956.1 | - | 17 - 18 | 2249 | 88 | ||
BC013502.1 | - | 10 - 11 | 1291 | 4223599 | 88 | |
BC058195.1 | - | 19 - 20 | 2489 | 5356348 | 88 | |
BP767442.1 | - | 1 - 2 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |