Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008968.3 | NP_032994.1 | 5 - 6 | 788 | 87 | |
GenBank mRNA | AB001607.1 | - | 5 - 6 | 706 | 87 | |
AK048059.1 | - | 841 | 87 | |||
AK076585.1 | - | 5 - 6 | 789 | 87 | ||
AY285779.1 | - | 1 - 2 | 137 | 87 | ||
BC062151.1 | - | 5 - 6 | 795 | 6512164 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |