Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252578.1 | NP_001239507.1 | 16 - 17 | 2677 | 72 | |
NM_001252579.1 | NP_001239508.1 | 16 - 17 | 2523 | 72 | ||
NM_028072.5 | NP_082348.2 | 16 - 17 | 2519 | 72 | ||
GenBank mRNA | AK008108.1 | - | 7 - 8 | 782 | 72 | |
AK028874.1 | - | 16 - 17 | 2434 | 72 | ||
AK034712.1 | - | 16 - 17 | 2504 | 72 | ||
AK036685.1 | - | 16 - 17 | 2455 | 72 | ||
AK049170.1 | - | 11 - 12 | 1434 | 72 | ||
AK081643.1 | - | 16 - 17 | 2477 | 72 | ||
AK129316.1 | - | 16 - 17 | 2677 | 72 | ||
AK133336.1 | - | 2493 | 72 | |||
AK165183.1 | - | 16 - 17 | 2431 | 72 | ||
AY101177.1 | - | 15 - 16 | 2338 | 72 | ||
BC062900.1 | - | 16 - 17 | 2475 | 72 | ||
BC141086.1 | - | 15 - 16 | 2272 | 9055698 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |