Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271962.1 | NP_001258891.1 | 2 - 3 | 166 | 108 | |
NM_001271963.1 | NP_001258892.1 | 2 - 3 | 342 | 108 | ||
NM_010165.3 | NP_034295.1 | 2 - 3 | 343 | 108 | ||
GenBank mRNA | AK137718.1 | - | 2 - 3 | 338 | 108 | |
AK141765.1 | - | 2 - 3 | 342 | 108 | ||
AY212511.1 | - | 1 - 2 | 103 | 108 | ||
AY212512.1 | - | 1 - 2 | 103 | 108 | ||
BC003755.1 | - | 2 - 3 | 343 | 3497245 | 108 | |
U61111.1 | - | 1 - 2 | 84 | 108 | ||
U71208.1 | - | 2 - 3 | 168 | 108 | ||
U81603.1 | - | 1 - 2 | 83 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |