Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199902.1 | NP_001186831.1 | 2 - 3 | 343 | 86 | |
GenBank mRNA | BC048510.1 | - | 2 - 3 | 127 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |