Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trhde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146241.2 | NP_666353.1 | 11 - 12 | 2201 | 103 | |
GenBank mRNA | AK029833.1 | - | 1531 | 103 | ||
AK034093.1 | - | 11 - 12 | 1580 | 103 | ||
BC032288.1 | - | 11 - 12 | 2206 | 5368580 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |