Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100029104.mm.1 |
NM_026068.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029104.mm.1 |
NM_026068.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029104.mm.1 |
TC1100003139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029106.mm.1 |
NM_026068.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029106.mm.1 |
NM_026068.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029106.mm.1 |
TC1100003139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029108.mm.1 |
NM_026068.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029108.mm.1 |
NM_026068.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029108.mm.1 |
TC1100003139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029111.mm.1 |
NM_026068.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029111.mm.1 |
NM_026068.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029111.mm.1 |
TC1100003139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029109.mm.1 |
NM_026068.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029109.mm.1 |
NM_026068.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029109.mm.1 |
TC1100003139.mm.1 |
MTA TC |