Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053245.2 | NP_444475.2 | 4 - 5 | 696 | 62 | |
GenBank mRNA | AF151392.1 | - | 565 | 62 | ||
AF296412.1 | - | 4 - 5 | 645 | 62 | ||
BC028285.1 | - | 5 - 6 | 774 | 4501401 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |