Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026810.2 | NP_081086.2 | 10 - 11 | 969 | 100 | |
GenBank mRNA | AF250844.1 | - | 10 - 11 | 885 | 100 | |
AK038265.1 | - | 10 - 11 | 962 | 100 | ||
AK145646.1 | - | 10 - 11 | 916 | 100 | ||
AK168849.1 | - | 10 - 11 | 908 | 100 | ||
BC021815.1 | - | 10 - 11 | 923 | 4020489 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |