Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081381.1 | NP_001074850.1 | 4 - 5 | 427 | 87 | |
GenBank mRNA | AK011270.1 | - | 5 - 6 | 583 | 87 | |
AK075986.1 | - | 4 - 5 | 424 | 87 | ||
BC048732.1 | - | 4 - 5 | 402 | 6512688 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |