Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177325.3 | NP_796299.2 | 2 - 3 | 242 | 56 | |
GenBank mRNA | AK129350.1 | - | 2 - 3 | 196 | 56 | |
AK134365.1 | - | 2 - 3 | 226 | 56 | ||
AK154063.1 | - | 2 - 3 | 229 | 56 | ||
AK165662.1 | - | 2 - 3 | 233 | 56 | ||
BC031531.1 | - | 2 - 3 | 215 | 4014819 | 56 | |
CD547366.1 | - | 2 - 3 | 242 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |