Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081164.1 | NP_001074633.1 | 8 - 9 | 913 | 72 | |
NM_001256033.1 | NP_001242962.1 | 8 - 9 | 913 | 72 | ||
GenBank mRNA | AK122429.1 | - | 3 - 4 | 204 | 72 | |
BC138373.1 | - | 8 - 9 | 885 | 8861393 | 72 | |
BC145259.1 | - | 8 - 9 | 885 | 9053812 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |