Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167680.1 | NP_001161152.1 | 14 - 15 | 1944 | 73 | |
NM_172572.3 | NP_766160.2 | 14 - 15 | 2158 | 73 | ||
GenBank mRNA | AK028872.1 | - | 14 - 15 | 2159 | 73 | |
AK154639.1 | - | 14 - 15 | 1924 | 73 | ||
AK171085.1 | - | 14 - 15 | 1940 | 73 | ||
AY167043.1 | - | 14 - 15 | 1944 | 73 | ||
BC031398.1 | - | 8 - 9 | 977 | 73 | ||
BC052182.1 | - | 14 - 15 | 1892 | 6335022 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |