Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079865.2 | NP_001073334.2 | 8 - 9 | 1170 | 135 | |
GenBank mRNA | AK040349.1 | - | 8 - 9 | 1172 | 135 | |
BC117742.1 | - | 8 - 9 | 1170 | 40090393 | 135 | |
BC117743.1 | - | 6 - 7 | 777 | 40090395 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |