Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144930.2 | NP_659179.1 | 5 - 6 | 728 | 66 | |
GenBank mRNA | AB023632.1 | - | 1 - 2 | 131 | 66 | |
AI786051.1 | - | 1 - 2 | 124 | 66 | ||
AK136491.1 | - | 5 - 6 | 720 | 66 | ||
BC013479.1 | - | 5 - 6 | 703 | 4239756 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |