Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195413.1 | NP_001182342.1 | 9 - 10 | 753 | 77 | |
NM_145601.1 | NP_663576.1 | 9 - 10 | 753 | 77 | ||
GenBank mRNA | BC016201.1 | - | 9 - 10 | 753 | 4527347 | 77 |
BC051415.1 | - | 7 - 8 | 1662 | 77 | ||
HQ116386.1 | - | 9 - 10 | 753 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |