Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100018695.mm.1 |
NM_198022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018695.mm.1 |
NM_198022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018695.mm.1 |
TC1100001965.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018696.mm.1 |
NM_198022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018696.mm.1 |
NM_198022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018696.mm.1 |
TC1100001965.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018702.mm.1 |
NM_198022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018702.mm.1 |
NM_198022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018702.mm.1 |
TC1100001965.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018699.mm.1 |
NM_198022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018699.mm.1 |
NM_198022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018699.mm.1 |
TC1100001965.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018701.mm.1 |
NM_198022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018701.mm.1 |
NM_198022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018701.mm.1 |
TC1100001965.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018703.mm.1 |
NM_198022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018703.mm.1 |
NM_198022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018703.mm.1 |
TC1100001965.mm.1 |
MTA TC |