Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018822.3 | NP_061292.2 | 6 - 7 | 767 | 101 | |
GenBank mRNA | AF153827.1 | - | 6 - 7 | 741 | 101 | |
AF304053.1 | - | 6 - 7 | 741 | 101 | ||
AF304054.1 | - | 6 - 7 | 741 | 101 | ||
AK052266.1 | - | 6 - 7 | 760 | 101 | ||
AK141266.1 | - | 6 - 7 | 715 | 101 | ||
BC130268.1 | - | 6 - 7 | 764 | 40143074 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |