Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195390.1 | NP_001182319.1 | 2 - 3 | 264 | 76 | |
GenBank mRNA | AK169037.1 | - | 2 - 3 | 211 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |