Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195390.1 | NP_001182319.1 | 7 - 8 | 607 | 128 | |
NM_133850.2 | NP_598611.2 | 6 - 7 | 532 | 128 | ||
GenBank mRNA | AK008447.1 | - | 6 - 7 | 487 | 128 | |
AK009274.1 | - | 6 - 7 | 507 | 128 | ||
AK077349.1 | - | 6 - 7 | 473 | 128 | ||
AK078774.1 | - | 6 - 7 | 491 | 128 | ||
AK089926.1 | - | 6 - 7 | 475 | 128 | ||
AK164805.1 | - | 6 - 7 | 441 | 128 | ||
AK169037.1 | - | 7 - 8 | 554 | 128 | ||
BC003310.1 | - | 6 - 7 | 465 | 128 | ||
BC065794.1 | - | 6 - 7 | 433 | 128 | ||
BC092215.1 | - | 6 - 7 | 449 | 6819383 | 128 | |
BC094216.1 | - | 6 - 7 | 489 | 6820900 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |