Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r1l-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005746.1 | - | 2 - 3 | 62 | 63 | |
AK029171.1 | - | 2 - 3 | 62 | 63 | ||
AK046224.1 | - | 2 - 3 | 65 | 63 | ||
AK161459.1 | - | 1 - 2 | 61 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |