Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008679.3 | NP_032705.2 | 15 - 16 | 3209 | 168 | |
GenBank mRNA | AK137517.1 | - | 4 - 5 | 880 | 168 | |
AK142306.1 | - | 15 - 16 | 3114 | 168 | ||
AK156345.1 | - | 15 - 16 | 3174 | 156 | ||
BC086482.1 | - | 15 - 16 | 3179 | 30104508 | 168 | |
BC093478.1 | - | 12 - 13 | 2840 | 168 | ||
BC141177.1 | - | 14 - 15 | 3025 | 9055789 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |