Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogfod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093757.1 | NP_001087226.1 | 5 - 6 | 653 | 75 | |
NM_177767.4 | NP_808435.3 | 5 - 6 | 653 | 75 | ||
GenBank mRNA | AK029629.1 | - | 5 - 6 | 655 | 75 | |
AK122532.1 | - | 5 - 6 | 557 | 75 | ||
AK156765.1 | - | 5 - 6 | 623 | 75 | ||
AK164189.1 | - | 5 - 6 | 540 | 75 | ||
AK166343.1 | - | 5 - 6 | 576 | 75 | ||
BC040267.1 | - | 5 - 6 | 613 | 5364674 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |