Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011787.2 | NP_035917.2 | 8 - 9 | 1259 | 110 | |
GenBank mRNA | AF124144.1 | - | 8 - 9 | 1467 | 110 | |
AK038442.1 | - | 8 - 9 | 1227 | 110 | ||
AK157327.1 | - | 8 - 9 | 1265 | 110 | ||
AK170716.1 | - | 8 - 9 | 1245 | 110 | ||
AK187742.1 | - | 2 - 3 | 143 | 110 | ||
AK190091.1 | - | 2 - 3 | 143 | 110 | ||
AK191669.1 | - | 2 - 3 | 143 | 110 | ||
BC034538.1 | - | 8 - 9 | 1217 | 4209258 | 110 | |
BC040338.1 | - | 8 - 9 | 1185 | 3710780 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |