Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cse1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023565.3 | NP_076054.1 | 15 - 16 | 1706 | 78 | |
GenBank mRNA | AF301152.1 | - | 15 - 16 | 1706 | 78 | |
AK138940.1 | - | 1 - 2 | 130 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |