Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144831.2 | NP_659080.2 | 2 - 3 | 320 | 126 | |
GenBank mRNA | AK078552.1 | - | 2 - 3 | 259 | 126 | |
AK142226.1 | - | 2 - 3 | 254 | 126 | ||
AK170616.1 | - | 2 - 3 | 247 | 126 | ||
AK172600.1 | - | 2 - 3 | 260 | 126 | ||
AK220485.1 | - | 2 - 3 | 279 | 126 | ||
BC065064.1 | - | 2 - 3 | 249 | 126 | ||
BC132445.1 | - | 2 - 3 | 304 | 40130722 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |