Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144831.2 | NP_659080.2 | 8 - 9 | 1372 | 85 | |
GenBank mRNA | AK078552.1 | - | 8 - 9 | 1311 | 85 | |
AK142226.1 | - | 8 - 9 | 1306 | 85 | ||
AK170616.1 | - | 8 - 9 | 1300 | 85 | ||
AK172600.1 | - | 8 - 9 | 1317 | 85 | ||
AK220485.1 | - | 8 - 9 | 1349 | 85 | ||
BC065064.1 | - | 8 - 9 | 1301 | 85 | ||
BC132445.1 | - | 8 - 9 | 1356 | 40130722 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |