Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085378.2 | NP_001078847.1 | 35 - 36 | 4878 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |