Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd6ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164677.1 | NP_001158149.1 | 15 - 16 | 2289 | 66 | |
NM_001164678.1 | NP_001158150.1 | 15 - 16 | 2274 | 66 | ||
NM_011052.2 | NP_035182.2 | 15 - 16 | 2274 | 66 | ||
GenBank mRNA | AF119955.1 | - | 15 - 16 | 2235 | 66 | |
AF176514.1 | - | 2 - 3 | 112 | 66 | ||
AJ005073.1 | - | 15 - 16 | 2122 | 66 | ||
AK029827.1 | - | 15 - 16 | 2241 | 66 | ||
AK031256.1 | - | 15 - 16 | 2275 | 66 | ||
AK033943.1 | - | 15 - 16 | 2263 | 66 | ||
AK129340.1 | - | 15 - 16 | 2244 | 66 | ||
AK145893.1 | - | 15 - 16 | 2262 | 66 | ||
AK147736.1 | - | 15 - 16 | 2247 | 66 | ||
AK167574.1 | - | 15 - 16 | 2232 | 66 | ||
AK169704.1 | - | 15 - 16 | 2275 | 66 | ||
AK188363.1 | - | 2 - 3 | 220 | 66 | ||
AK208374.1 | - | 3 - 4 | 296 | 66 | ||
BC002261.1 | - | 2 - 3 | 223 | 66 | ||
BC026823.1 | - | 15 - 16 | 2216 | 4038684 | 66 | |
BC051123.1 | - | 15 - 16 | 2125 | 6529850 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |