Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018533.1 | - | 7 - 8 | 1034 | 93 | |
AK036488.1 | - | 7 - 8 | 1034 | 93 | ||
AK137450.1 | - | 7 - 8 | 1569 | 93 | ||
AK146004.1 | - | 7 - 8 | 1034 | 93 | ||
AK150320.1 | - | 7 - 8 | 1035 | 93 | ||
AK150370.1 | - | 7 - 8 | 1034 | 93 | ||
AK151324.1 | - | 7 - 8 | 1034 | 93 | ||
AK151480.1 | - | 7 - 8 | 1035 | 93 | ||
AK152477.1 | - | 7 - 8 | 1034 | 93 | ||
AK152906.1 | - | 7 - 8 | 1034 | 93 | ||
AK153068.1 | - | 7 - 8 | 1034 | 93 | ||
AK153166.1 | - | 7 - 8 | 1034 | 93 | ||
AK153395.1 | - | 7 - 8 | 1034 | 93 | ||
AK154662.1 | - | 7 - 8 | 1034 | 93 | ||
AK156481.1 | - | 7 - 8 | 1034 | 93 | ||
AK157147.1 | - | 7 - 8 | 1034 | 93 | ||
AK157170.1 | - | 7 - 8 | 1034 | 93 | ||
AK157249.1 | - | 7 - 8 | 1034 | 93 | ||
AK157700.1 | - | 7 - 8 | 1033 | 93 | ||
AK159835.1 | - | 7 - 8 | 1034 | 93 | ||
BC005450.1 | - | 7 - 8 | 1015 | 3487214 | 93 | |
CT010225.1 | - | 6 - 7 | 980 | 93 | ||
M32489.1 | - | 7 - 8 | 1000 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |