Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028312.3 | NP_082588.2 | 1 - 2 | 159 | 67 | |
GenBank mRNA | AK012610.1 | - | 1 - 2 | 160 | 67 | |
BC026668.1 | - | 1 - 2 | 109 | 5006213 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |