Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170684.2 | NP_733785.1 | 5 - 6 | 711 | 70 | |
GenBank mRNA | AK049823.1 | - | 5 - 6 | 640 | 70 | |
AK162900.1 | - | 5 - 6 | 712 | 70 | ||
AU079878.1 | - | 5 - 6 | 630 | 70 | ||
BC119328.1 | - | 5 - 6 | 610 | 8734077 | 70 | |
BC137595.1 | - | 5 - 6 | 610 | 8860614 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |