Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029360.3 | NP_083636.2 | 2 - 3 | 313 | 65 | |
GenBank mRNA | AK008064.1 | - | 2 - 3 | 311 | 65 | |
BB869909.1 | - | 2 - 3 | 314 | 65 | ||
BC010782.1 | - | 2 - 3 | 273 | 4219239 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |