Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029360.3 | NP_083636.2 | 3 - 4 | 449 | 93 | |
GenBank mRNA | AK008064.1 | - | 3 - 4 | 447 | 93 | |
BC010782.1 | - | 3 - 4 | 409 | 4219239 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |