Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145404.1 | NP_663379.1 | 2 - 3 | 220 | 110 | |
GenBank mRNA | AK040103.1 | - | 2 - 3 | 252 | 110 | |
AK154255.1 | - | 2 - 3 | 247 | 110 | ||
AK173304.1 | - | 2 - 3 | 240 | 110 | ||
AY673972.1 | - | 1 - 2 | 90 | 110 | ||
BC006705.1 | - | 2 - 3 | 220 | 3583763 | 110 | |
BC012845.1 | - | 1 - 2 | 41 | 110 | ||
BC057177.1 | - | 2 - 3 | 244 | 5010547 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |