Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a6os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007567.2 | NP_001007568.1 | 3 - 4 | 691 | 112 | |
GenBank mRNA | AK146733.1 | - | 3 - 4 | 692 | 112 | |
BC054118.1 | - | 3 - 4 | 689 | 30021749 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |