Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139516.1 | NP_001132988.1 | 11 - 12 | 1544 | 83 | |
NM_011249.2 | NP_035379.2 | 11 - 12 | 1544 | 83 | ||
GenBank mRNA | AK033402.1 | - | 11 - 12 | 1538 | 83 | |
AK147064.1 | - | 11 - 12 | 1545 | 83 | ||
AK156055.1 | - | 11 - 12 | 1523 | 83 | ||
AK214366.1 | - | 2 - 3 | 236 | 83 | ||
BC060124.1 | - | 11 - 12 | 1511 | 6408844 | 83 | |
BC069179.1 | - | 3 - 4 | 322 | 83 | ||
U27177.1 | - | 11 - 12 | 1528 | 83 | ||
U27178.1 | - | 11 - 12 | 1528 | 83 | ||
U33320.1 | - | 11 - 12 | 1511 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |