Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025853.3 | NP_080129.2 | 5 - 6 | 599 | 129 | |
GenBank mRNA | AK006246.1 | - | 5 - 6 | 600 | 129 | |
AK017740.1 | - | 5 - 6 | 600 | 129 | ||
AK017813.1 | - | 5 - 6 | 554 | 129 | ||
AK139887.1 | - | 5 - 6 | 1169 | 129 | ||
AK145373.1 | - | 6 - 7 | 427 | 129 | ||
BC025079.1 | - | 5 - 6 | 582 | 3499844 | 129 | |
BC046807.1 | - | 5 - 6 | 560 | 6829342 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |