Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139520.1 | NP_001132992.1 | 9 - 10 | 1226 | 70 | |
NM_018851.3 | NP_061339.3 | 9 - 10 | 1226 | 70 | ||
GenBank mRNA | AK054490.1 | - | 9 - 10 | 1125 | 70 | |
AK078936.1 | - | 9 - 10 | 1169 | 70 | ||
AK128954.1 | - | 8 - 9 | 1024 | 70 | ||
AK149865.1 | - | 9 - 10 | 1101 | 70 | ||
AK151335.1 | - | 9 - 10 | 1100 | 70 | ||
AK153390.1 | - | 9 - 10 | 1100 | 70 | ||
AK170688.1 | - | 9 - 10 | 1125 | 70 | ||
AK171565.1 | - | 9 - 10 | 1126 | 70 | ||
BC012721.1 | - | 9 - 10 | 1163 | 4037046 | 70 | |
BC067198.1 | - | 9 - 10 | 1149 | 6826682 | 70 | |
U15635.1 | - | 9 - 10 | 1150 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |