Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109752.1 | NP_001103222.1 | 5 - 6 | 639 | 61 | |
NM_007864.3 | NP_031890.1 | 5 - 6 | 648 | 61 | ||
GenBank mRNA | BC014807.1 | - | 5 - 6 | 384 | 4501403 | 61 |
BI738861.1 | - | 5 - 6 | 393 | 61 | ||
CA320223.1 | - | 5 - 6 | 639 | 61 | ||
D50621.1 | - | 5 - 6 | 390 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |