Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800021401.mm.1 |
NM_181990.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021401.mm.1 |
NM_181990.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021401.mm.1 |
TC0800002798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800021402.mm.1 |
NM_181990.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021402.mm.1 |
NM_181990.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021402.mm.1 |
TC0800002798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800021404.mm.1 |
NM_181990.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021404.mm.1 |
NM_181990.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021404.mm.1 |
TC0800002798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800021405.mm.1 |
NM_181990.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021405.mm.1 |
NM_181990.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021405.mm.1 |
TC0800002798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800021395.mm.1 |
NM_181990.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021395.mm.1 |
NM_181990.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021395.mm.1 |
TC0800002798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800021400.mm.1 |
NM_181990.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021400.mm.1 |
NM_181990.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021400.mm.1 |
TC0800002798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800021396.mm.1 |
NM_181990.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021396.mm.1 |
NM_181990.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800021396.mm.1 |
TC0800002798.mm.1 |
MTA TC |