Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197999.2 | NP_932116.2 | 10 - 11 | 1495 | 93 | |
GenBank mRNA | BC024548.1 | - | 2 - 3 | 247 | 93 | |
BC026641.1 | - | 2 - 3 | 251 | 93 | ||
BC027185.1 | - | 10 - 11 | 1473 | 4194872 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |