Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033371.3 | NP_001028543.1 | 7 - 8 | 1345 | 85 | |
NM_001170788.1 | NP_001164259.1 | 10 - 11 | 1674 | 85 | ||
NM_001170789.1 | NP_001164260.1 | 7 - 8 | 1351 | 85 | ||
GenBank mRNA | AK133042.1 | - | 7 - 8 | 1345 | 85 | |
BC036564.1 | - | 5 - 6 | 962 | 85 | ||
BC150957.1 | - | 7 - 8 | 1337 | 9087868 | 85 | |
BC172120.1 | - | 7 - 8 | 1343 | 9054406 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |