Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033371.3 | NP_001028543.1 | 8 - 9 | 1500 | 64 | |
NM_001170788.1 | NP_001164259.1 | 11 - 12 | 1829 | 64 | ||
NM_001170789.1 | NP_001164260.1 | 8 - 9 | 1506 | 64 | ||
GenBank mRNA | AK133042.1 | - | 8 - 9 | 1500 | 64 | |
BC036564.1 | - | 6 - 7 | 1117 | 64 | ||
BC150957.1 | - | 8 - 9 | 1492 | 9087868 | 64 | |
BC172120.1 | - | 8 - 9 | 1498 | 9054406 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |