Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133702.2 | NP_598463.2 | 13 - 14 | 1647 | 162 | |
GenBank mRNA | AK078524.1 | - | 13 - 14 | 1538 | 162 | |
AK157743.1 | - | 13 - 14 | 1587 | 162 | ||
BC004785.1 | - | 8 - 9 | 926 | 162 | ||
BC024874.1 | - | 13 - 14 | 1526 | 3984027 | 162 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |