Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200041721.mm.1 |
NM_001159376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041721.mm.1 |
NM_001159376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041721.mm.1 |
TC0200004920.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041716.mm.1 |
NM_001159376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041716.mm.1 |
NM_001159376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041716.mm.1 |
NM_145744.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041716.mm.1 |
NM_145744.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041716.mm.1 |
TC0200004920.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041718.mm.1 |
NM_001159376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041718.mm.1 |
NM_001159376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041718.mm.1 |
NM_145744.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041718.mm.1 |
NM_145744.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041718.mm.1 |
TC0200004920.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041720.mm.1 |
NM_001159376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041720.mm.1 |
NM_001159376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041720.mm.1 |
TC0200004920.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041717.mm.1 |
NM_001159376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041717.mm.1 |
NM_001159376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041717.mm.1 |
NM_145744.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041717.mm.1 |
NM_145744.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041717.mm.1 |
TC0200004920.mm.1 |
MTA TC |