Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025876.2 | NP_080152.1 | 1 - 2 | 39 | 64 | |
GenBank mRNA | AK010081.1 | - | 1 - 2 | 39 | 64 | |
AK010097.1 | - | 1 - 2 | 39 | 64 | ||
AK088517.1 | - | 1 - 2 | 48 | 64 | ||
AK136570.1 | - | 1 - 2 | 52 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |