Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081151.1 | NP_001074620.1 | 3 - 4 | 640 | 63 | |
GenBank mRNA | AK039455.1 | - | 3 - 4 | 639 | 63 | |
BC112432.1 | - | 3 - 4 | 639 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |