Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010911.2 | NP_035041.2 | 8 - 9 | 1003 | 77 | |
GenBank mRNA | AJ222660.1 | - | 8 - 9 | 1002 | 77 | |
AK005724.1 | - | 8 - 9 | 993 | 77 | ||
AK075575.1 | - | 8 - 9 | 1001 | 77 | ||
AK132838.1 | - | 8 - 9 | 1004 | 77 | ||
AK148527.1 | - | 8 - 9 | 1004 | 77 | ||
AK159828.1 | - | 8 - 9 | 992 | 77 | ||
BC010586.1 | - | 8 - 9 | 981 | 3711352 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |